Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: KPL
5260-0060
95058-916EA 275.03 USD
95058-916
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Secondary Goat Anti-IgG(H+L) Reacts with Rabbit (Lapine)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR