Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: KPL
5220-0371
95058-796EA 211.18 USD
95058-796
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Hamster

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR