PERMA-HAND™ Silk Sutures, ETHICON, National Distribution and Contracting
Supplier: Healthcare Products
ETH 683G ETH 632G ETH 706G ETH N267H ETH K952H ETH K891H ETH K882H ETH K833H ETH 737G ETH 675H ETH 624H ETH 443H ETH K571H ETH 736G ETH 768G ETH A182H ETH SA64H ETH LA53G ETH LA54G ETH LA55G ETH K831H ETH K834H ETH 734G ETH M104T ETH N266H ETH 640G ETH 1679H ETH K802H ETH K889H ETH K890H ETH K880H ETH 707G ETH K872H ETH 678G ETH 682G ETH K809H ETH 7734G ETH 677G ETH K873H ETH K870H ETH A185H ETH 685G ETH 639G
95056-870BX 211.25 USD
95056-870 95056-896 95056-912 95056-956 95056-994 95056-996 95056-998 95057-004 95057-054 95057-058 95057-070 95057-088 95057-090 95056-914 95056-916 95056-922 95056-960 95056-988 95056-990 95056-992 95057-002 95057-006 95057-052 95057-094 95057-096 95056-872 95056-890 95056-948 95056-952 95056-954 95056-968 95056-972 95057-000 95057-062 95057-064 95057-066 95057-092 95057-048 95057-060 95056-950 95056-966 95057-034 95057-068 95057-086
PERMA-HAND™ Silk Sutures, ETHICON, National Distribution and Contracting
Sutures
Nonabsorbable, sterile, surgical sutures are composed of an organic protein called fibroin, which is derived from the domesticated species Bombyx mori (B
mori) of the family Bombycidae. Indicated for use in general soft tissue approximation and ligation, including use in cardiovascular, ophthalmic and neurological procedures. Contraindicated in patients with known sensitivities or allergies to silk. Not recommended where permanent retention of tensile strength is required. Sutures are processed to remove the natural waxes and gums, dyed black, and coated with a special wax mixture.