Coated VICRYL® (polyglactin 910) Sutures, ETHICON, National Distribution and Contracting
Supplier: Healthcare Products
ETH J546G ETH J451H ETH J311H ETH J214H ETH J409G ETH J392H ETH J339H ETH J488G ETH J494G ETH J845G ETH J869H ETH J386H ETH J310H ETH J340H ETH J317H ETH J463G ETH J844G ETH J495G ETH J315H ETH J338H ETH J316H ETH J337H
95057-008BX 1881.07 USD
95057-008 95057-012 95057-022 95057-028 95057-132 95056-874 95056-928 95056-938 95056-940 95056-944 95056-946 95057-014 95057-020 95057-098 95057-126 95057-136 95056-936 95056-942 95057-010 95057-016 95057-024 95057-026 95057-030 95057-134 95056-926 95056-876 95057-018
Coated VICRYL® (polyglactin 910) Sutures, ETHICON, National Distribution and Contracting
Sutures
Absorbable sterile synthetic sutures feature exclusive coating technology for smooth passage through tissue, ease of knot tying, unsurpassed knot-sliding capabilities, and virtually no package memory for easier handling
Indicated for use in general soft tissue approximation and ligation, including use in ophthalmic and orthopedic procedures, but not for use in cardiovascular or neurological tissues. Not intended for use where extended approximation of tissue is required.
Información para pedidos: Each suture comes with one or two needles attached.