PD MultiTrap™ G-25, Filter Plates, 96-Well, Cytiva
Supplier: Cytiva
28-9180-06
95056-248PK 1429 USD
95056-248
PD MultiTrap™ G-25, Filter Plates, 96-Well, Cytiva
Microplates Filter Plates
PD MultiTrap G-25 is designed for high-throughput desalting and buffer exchange of multiple samples.
  • Convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr) with high reproducibility well-to-well and plate-to-plate
  • Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds
  • High desalting capacity
  • Suitable for manual use as well as for automation together with robotic systems
  • Sample preparation in the range of 70 to 130 μl without sample dilution
PD MultiTrap G-25 isMore Product Information