Instrument Prep Enzyme Foam, Integra™ Miltex®
Supplier: Miltex
3-760
95039-236CS 450.88 USD
95039-236
Instrument Prep Enzyme Foam, Integra™ Miltex®
Cleaning Agents Detergents and Surface Cleansers
Ready-to-use foaming spray for pre-cleaning of soiled instruments and scopes
Ideal for applications where instrument cleaning is not immediately available. Simply spray on instruments until ready for cleaning. Prevents bio-burden from drying onto instruments prior to decontamination. Active enzymes work to break down blood, mucus, and other bio-burden for reduced cleaning time. No water dilution is required, preventing spills and splashing during transport. Easy and safe to use on metals, plastic, and rubber materials.