Disposable Biopsy Punches, Integra™ Miltex®
Supplier: Miltex
33-31 33-32 33-34 33-35 33-36 33-37 33-33 33-31B 33-31A 33-31AA 33-31A-P/25 33-34-P/25 33-31-P/25 33-38 33-31AA-P/25 33-32-P/25
21909-132PK 299.48 USD
21909-132 21909-136 21909-140 21909-142 21909-144 21909-146 21909-138 95039-096 95039-088 95039-090 95039-094 95039-102 95039-098 95039-104 95039-092 95039-100
Disposable Biopsy Punches, Integra™ Miltex®
Biopsy Punches
These biopsy punches offer the convenience of single-use disposability without sacrificing quality, feel, or performance
All punches feature seamless, stainless-steel, razor-sharp edges to eliminate torn or pulled biopsies. Ribbed handles provide comfort and control. Punches are color-coded with sizes clearly marked. Sterile, five-year shelf life.
Plunger-system models are available in five sizes and offer the added benefit of effortless ejection of lodged speciments. Non-plunger-system models are available in ten sizes for a wide range of procedures.
Información para pedidos: More Product Information