Thiamine hydrochloride ≥98% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

TH116-25KGBL TH116-100GM TH116-1KG TH116-25GM TH116-500GM
700009-328EA 4771.95 USD
700009-328 95036-270 95036-272 95036-274 95036-276
Thiamine hydrochloride ≥98% USP
Thiamine hydrochloride

Thiamine Hydrochloride, USP is the hydrochloride salt form of a material also known as vitamin B1. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Melting Pt: 252 °C
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR