Mini Dialysis Kit, Cytiva
Supplier: Cytiva
80-6483-75 80-6483-94 80-6484-13 80-6484-32
95017-768EA 532.6 USD
95017-768 95017-770 95017-772 95017-774
Mini Dialysis Kit, Cytiva
Tubes Dialysis Tubes
The disposable tubes in mini dialysis kit offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
  • Designed for efficient dialysis of small sample volumes
  • Uses disposable dialysis tubes with dialysis membrane incorporated into caps
  • Conical tube bottom maximizes sample recovery
Dialyzing small samples can be difficult and inefficient when the sample must be transferred into and out of dialysis bags and centrifuge tubes. The disposableMore Product Information