EAH Sepharose™ 4B Affinity Coupling Media, Cytiva
Supplier: Cytiva
17-0569-01
95016-888EA 701.73 USD
95016-888
EAH Sepharose™ 4B Affinity Coupling Media, Cytiva
Protein Purification Protein Purification Resins
EAH Sepharose™ 4B allows for the stable carbodiimide-based coupling of carboxyl groups to Sepharose® 4B via an 11-atom hydrophilic spacer arm that permits extremely stable coupling.
Stable carbodiimide-based coupling of carboxyl groups to 4B via an 11-atom hydrophilic spacer arm permits very stable coupling. Stable to all commonly used aqueous Sepharose buffers.