Barbed Elbows, Eldon James
Supplier: Avantor
94025-492PK 140.21 USD
94025-492 94025-494 94025-496 94025-498 94025-500 94025-502 94025-504 94025-506 94025-508 94025-510 94025-512 94025-514 94025-516 94025-518 94025-520 94025-522 94025-524 94025-526 94025-528 94025-530 94025-532 94025-534 94025-536 94025-538 94025-540 94025-542 94025-544 94025-546 94025-548 94025-550 94025-552 94025-554 94025-558 94025-566
Barbed Elbows, Eldon James
Fittings and Connectors
These elbows are made of Natural Kynar® material, which has a temperature range of -62°C to 107°C (-80 to 226°F), and a pressure of 260 psi at 21°C (70°F)
This material can be autoclaved to 140°C (285°F) and meets USP Class VI requirements. Generally, chemically resistant to most acids, salts, weak bases, halogenated solvents, alcohols, nuclear and UV radiation, oxidants, ozone and deionized water.
Certificaciones: RoHS Compliant.