BD BBL™ Nocardia ID Prepared Media QUAD Plates, BD Diagnostics
Supplier: BD
298309
90008-752PK 221.28 USD
90008-752
BD BBL™ Nocardia ID Prepared Media QUAD Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Prepared plated media are designed for the isolation of microorganisms from samples or specimens.
  • Supplied in QUAD plates
  • Each section contains a different media formulation
  • Ready for immediate use
These media combinations are supplied in QUAD plates, which are 100x15 mm petri-style dishes divided into four sections. Each section contains a different media formulation. Plates are ready for immediate use. Store at 2 to 8°C (36 to 46°F).