BBL™ Inhibitory Mold Agar, BD Biosciences
Supplier: BD
292846
90004-442EA 290.56 USD
90004-442
BBL™ Inhibitory Mold Agar, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Inhibitory mold agar is used for the isolation of pathogenic fungi.
  • Manufactured in an animal origin-free, antibiotic-free facility
  • Mycoplasma testing upon release
  • Endotoxin testing upon release
The individual ingredients and final dehydrated product are carefully quality controlled to assure consistent, high-quality performance and obviate the need for media to be prepared in the laboratory from raw materials.