Anaerobic Systems, BD Diagnostics
Supplier: BD
260411 260467 260608 260610 260614 260638 270124 260414 260607 260609 260619 260463 260413 260618 260637
90003-564EA 942.96 USD
90003-564 90003-582 90003-586 90003-590 90003-592 90003-602 90003-604 90003-608 90003-614 90003-822 BB4360414 90003-584 90003-588 90003-596 90003-580 90003-606 BB4360413 90003-594 90003-612
Anaerobic Systems, BD Diagnostics
Anaerobic Systems
Designed for the generation of anearobic, microaerophilic (reduced oxygen), or CO2-enriched environments.
The BD GasPak™ 100 and 150 self-contained anaerobic systems consist of one polycarbonate jar (vented or non-vented), lid with O-ring gasket, catalyst reaction chamber, BBL room temperature catalyst charges, one rack, and one tube holder. The BD GasPak™ 100 system features two catalyst charges; the BD GasPak™ 150 system features ten charges. These systems use hydrogen and carbon dioxide generator envelopes and do not require a vacuum pump, gas tank, manometer, or pressure reducingMore Product Information