BD Difco™ Salmonella Antiserum O Vi, BD Diagnstics
Supplier: BD
228271
90001-986EA 268.6 USD
90001-986
BD Difco™ Salmonella Antiserum O Vi, BD Diagnstics
Microbiology Test Systems Microorganism Tests
BD offers antigens and antisera that can facilitate diagnosis of several diseases and meet your microbiology needs.
  • Used in slide agglutination tests for the identification of Salmonella Vi
BD Difco™ Salmonella Vi Antisera are lyophilized, polyclonal rabbit antisera containing approximately 0.04% Thimerosal as a preservative. When properly rehydrated and used as recommended, each vial of BD Difco™ Salmonella Vi Antisera contains sufficient reagent for 60 tests. BD Difco™ Salmonella Vi Antisera are prepared with representative strains of these serogroups and are not absorbed.