Difco™ Purple Agar Base, BD
Supplier: BD
222810
90001-708EA 372.47 USD
90001-708
Difco™ Purple Agar Base, BD
Media for Microbiology Dehydrated Media for Microbiology
Purple Agar Base and Purple Broth Base are used with added carbohydrate in differentiating pure cultures of bacteria.
  • Carbohydrate-free peptone with the pH indicator bromcresol purple
  • Indicator changes with the amount of acid produced
  • No reaction means no fermentation
Purple Agar Base is used primarily for the differentiation and presumptive identification of gram-negative enteric bacilli based on patterns of carbohydrate fermentation.
Order Now
SPECIFICATIONS
Environmentally Preferable