VWR® Forced Air Microbiological Incubators
Supplier: VWR International
89511-424 89511-426 89511-428 89511-430 10144-890 10144-892 10144-894 10144-896 10145-406 10145-408 10145-410
89511-424EA 3441.08 USD
89511-424 89511-426 89511-428 89511-430 10144-890 10144-892 10144-894 10144-896 10145-406 10145-408 10145-410
VWR® Forced Air Microbiological Incubators
Incubators Standard Incubators
VWR® Forced Air incubators provide an even higher temperature uniformity and stability for reproducible results in microbiology and research labs.
  • Outstanding temperature stability and uniformity
  • Protects your valuable samples
  • Extremely versatile with a wide temperature range, Ambient +5 °C
  • Three benchtop sizes and one floor standing size ranging from 2.3 – 13.4 cu. ft.
  • 2 perforated stainless steel shelves included (55 lbs. capacity each; 66 lbs. in floor standing model)
  • Standard access port for monitoring and use of electrical equipment inside unit
  • Same
More Product Information