Cardboard Microscope Slide Mailers, Globe Scientific
Supplier: Globe Scientific
513001 513002 513003 513004 513025
89510-756CS 327.52 USD
89510-756 89510-758 89510-760 89510-762 89510-764
Cardboard Microscope Slide Mailers, Globe Scientific
Slide Mailers
These flat cardboard slide mailers securely hold slides with or without a cover glass
The 20-place cardboard slide holder contains a numerical index that corresponds to the inner slide compartments. The inside flaps are recessed so that there is no contact or contamination between the slides and the cover when closed. With a 360° hinge, the cover folds flat underneath to save space when the holder is in use.
They feature an inner finger groove for effortless slide removal along with a cover that opens for viewing. The flat surface area is ideal for labeling or writing any necessaryMore Product Information