Arium® Advance RO Type 3 Water Systems
Supplier: Sartorius
H2O-RO-1-T H2O-RO-1-B H2O-RO-2-B H2O-RO-2-T H2O-RO-3-B H2O-RO-3-T
89510-654EA 7314.97 USD
89510-654 89510-656 89510-658 89510-660 89510-662 89510-664
Arium® Advance RO Type 3 Water Systems
Water Purification Systems Pure and Ultra Pure Water Systems
The arium advance RO delivers Type 3 reverse osmosis water at rates of up to 16 L per hour.
  • Unique Bagtank Technology
  • Dispense Gun Connections
  • Touch-Screen Control Panel
  • Innovative iJust Technology
  • Sixteen Liters of Type 3 Reverse Osmosis Water per Hour
The system uses unique bagtank technology with dispense gun connections
Features include a touch-screen control panel and innovative iJust technology.
It attains the highest retention rates of ions along with optimal water yield and removes oxidants, heavy metal ions, and particulates from feedMore Product Information