VWR® Vacuum Ovens
Supplier: VWR International
89508-424 89508-426 89508-428 10144-910 10144-916 10144-918 10145-424 10145-426 10145-428
89508-424EA 7011.04 USD
89508-424 89508-426 89508-428 10144-910 10144-916 10144-918 10145-424 10145-426 10145-428
VWR® Vacuum Ovens
Ovens Vacuum Ovens
The 12.5 L unit features a top-mounted independent evacuation and venting vacuum fittings.
  • Heating system optimizes chamber space for drying, curing, vacuum embedding, and plating applications
  • Polycarbonate safety shield protects door glass
  • Built-in over-temperature protection
  • Vacuum level is displayed on gauges from 0 to 30 in Hg
  • Corrosion-resistant stainless-steel tubing for long-term performance
  • With two removable stacking aluminum shelves
  • Easy-to-clean Type 304 stainless-steel interior chamber
  • Powder-coated heavy-gauge steel exterior for durability
  • Two-
More Product Information