Ro-Con® Transport Square Fibre Drum, Greif
Supplier: Greif Brothers
FRT8039
40300-184EA 29.06 USD
40300-184 89501-592
Ro-Con® Transport Square Fibre Drum, Greif
Barrels, Kegs, Drums
For filling, handling, warehousing, and/or shipping of dry chemical products. Drum is constructed of convolutedly wound paperboard and features rounded corners, convex sides, sturdy single wall construction, telescoping cover, and offset tape closure.