C-MAG HP 7 IKATHERM® Hot Plate, IKA® Works
Supplier: IKA Works
0003581801 3581800
89495-340EA 1518.35 USD
89495-340 89495-342
C-MAG HP 7 IKATHERM® Hot Plate, IKA® Works
Heaters Hot Plates
Newly designed hotplate constructed of ceramic glass to offer excellent chemical resistance.
Standard features include a fixed safety circuit of 550 °C, Hot Top indicator for hot surface warning to prevent burns, exact temperature setting via digital display (LED), a digital error code display, and an elevated control panel for protection against liquid leaks. Also provided is a bushing according to DIN 12878 for connecting a contact thermometer (see 89028-114 series) to enable precise temperature control.
Units meet IP 21 protection class according to DIN EN 60529.
Certificaciones:More Product Information