Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Supplier: Biotium

20230-1MG
89493-370EA 266.7 USD
89493-370
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 555
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR