PTFE Covered Stainless Steel Stir Shaft with Retreat Curve Agitator, 10 mm, Ace Glass
Supplier: Ace Glass
13850-01 13850-04
89428-020EA 652.65 USD
89428-020 89428-022
PTFE Covered Stainless Steel Stir Shaft with Retreat Curve Agitator, 10 mm, Ace Glass
Stirrers Stirring Shafts
This PTFE covered stainless steel shaft features a solid PTFE retreat curve agitator
Featuring a retreat angle of 30 degrees and, for a better grip by the stir motor chuck, 50mm of exposed stainless steel shaft.