Vacuum Filter Tube, with Inner Standard Taper Joints on Top and Bottom, Ace Glass Incorporated
Supplier: Ace Glass
7827-01 7827-03 7827-05 7827-07
89428-000EA 256 USD
89428-000 89428-002 89428-004 89428-006
Vacuum Filter Tube, with Inner Standard Taper Joints on Top and Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This borosilicate glass Schlenk filter tube features an integral glass filter disc at the tube's mid-line, and inner joints at top and bottom.