Economy Series Cryo-Compact Circulators, JULABO
Supplier: JULABO
9400330.2 9400340.2
89421-668EA 4683.21 USD
89421-668 89421-670 89421-672 89421-674 89421-676 89421-678 89421-680 89421-682 89421-684 89421-752 89421-754
Economy Series Cryo-Compact Circulators, JULABO
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Cryo-Compact circulators provide a wide temperature range for laboratory applications in a space saving design.
  • German quality and engineering
  • Stainless steel construction
  • Wide temperature range
  • Small footprint
JULABO Economy Series Cryo-Compact Refrigerated and Heating Circulators are suitable for temperature control applications to external systems.
The Cryo-Compact refrigerated/heating circulators have a very small footprint which is ideal for placing on bench top or in a fume hood with limited space. The CF30 and CF40 models feature pumpMore Product Information