Anti-TACE Rabbit Polyclonal Antibody
Supplier: Prosci
1131
89415-234EA 567.31 USD
89415-234
Anti-TACE Rabbit Polyclonal Antibody
Antibodies
TACE Antibody: Tumor-necrosis factor-alpha is a proinflammatory cytokine and contributes to a variety of inflammatory disease responses and programmed cell death. TNF-alpha is synthesized as a 26K type II membrane-bound precursor that is cleaved by a convertase to generate secreted 17K mature TNF-alpha. TNF-alpha converting enzyme (TACE) protein was recently purified and the human and mouse TACE cDNAs were cloned by several groups separately. TACE is a membrane-bound metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease). TACE also processes otherMore Product Information