SHEL LAB Drosophila Refrigerated Peltier Incubator, Sheldon
Supplier: Sheldon Manufacturing
SRI20PF SRI3P SRI6PF
89409-362EA 16539.65 USD
89409-362 76458-746 76458-748
SHEL LAB Drosophila Refrigerated Peltier Incubator, Sheldon
Incubators Cooled Incubators
Using innovative peltier cooling technology, drosphila incubators offer a 78% reduction in energy consumption over traditional compressor based refrigerated incubators.
  • Robust design
  • Whisper-quiet operation
  • Over-temperature thermostat and alarm
These incubators are equipped with LED Lighting and Day/Night Controls to create ideal conditions for fruit flies
Independent over-temperature thermostat and alarm.
The robust cabinet design includes a mechanical convection system to provide improved temperature uniformity, digital temperature controllerMore Product Information