Lab Armor® Single Bead Blocks, SHEL LAB
Supplier: Sheldon Manufacturing
52100-BLK 52100-BLU 52100-GLD 52100-SLV
89409-268EA 107.81 USD
89409-268 89409-270 89409-272 89409-274 89409-276
Lab Armor® Single Bead Blocks, SHEL LAB
Block Heaters Blocks
The Bead Block™ is an aluminum block filled with 0.25L of Lab Armor Beads™ designed for use with any standard, commercially available dry bath
The Bead Block™ replaces common drilled-out aluminum heat blocks and provides optimal temperature uniformity and accuracy while incubating common lab vessels. These blocks accommodate a wide assortment of vessels, including microfuge tubes, test tubes, and even microplates and slides. The Bead Block™ eliminates the need for using different size blocks to fit different sample vessels and can be easily lifted out of the dry bath unit for portable useMore Product Information