Cell culture growth supplement, Trace Elements A, Corning®
Supplier: Corning
25-021-CI
89408-312EA 73.47 USD
89408-312
Cell culture growth supplement, Trace Elements A, Corning®
Cell Culture Supplements and Additives
Trace Elements A provides copper, zinc, iron, and selenium
The combination of Trace Elements A contains the trace elements of MCDB 104 and of MCDB 301.
Selenium aids in the detoxification of free radicals as a cofactor for GSH synthetase, while iron, copper, and zinc may be bound by serum protein.