HardyVal™ CSP Medium Complexity Kit, Basic, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
HVM2
89408-158EA 168.67 USD
89408-158
HardyVal™ CSP Medium Complexity Kit, Basic, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
Compliance made easy to verify the efficacy of aseptic technique.
  • The product is quality control tested for growth promotion, pH, and sterility
The Medium Complexity Level Kit, Basic is recommended for simulating medium risk level compounding manipulations and verification of aseptic transfer techniques of multiple solutions. Kit performs one complete aseptic technique verification challenge test for one technician.
Información de suministro: Each Kit Contains:
Tryptic Soy Broth (TSB), 50 ml Needle Port Septum Serum Vial (3 vials)
Sterile 50 ml Serum Vials, empty (6 vials)
Sterile 20 ml Serum Vials, empty (3 vials)
Whirl-Pak™ Bag (1 bag)
Results Log Sheet (1 sheet)