MacConkey Agar
Supplier: HARDY DIAGNOSTICS
G35BX G35 G36 Q63
89407-270CS 188.63 USD
89407-270 89407-268 89407-272 89407-274 76019-244
MacConkey Agar
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and differentiation of Gram negative bacteria, especially the Enterobacteriaceae group.