Tygon S3™ B-44-4X Food and Beverage Dispensing Tubing
Supplier: Saint Gobain Life Sciences
AAA00065 AAA00001 AAA00002 AAA00004 AAA00006 AAA00009 AAA00010 AAA00011 AAA00018 AAA00023 AAA00028 AAA00029 AAA00036 AAA00053 AAA00062 AAA00064
89404-196PK 349.74 USD
89404-196 89404-216 89404-224 89404-226 89404-232 89404-238 89404-240 89404-156 89404-158 89404-160 89404-162 89404-164 89404-166 89404-168 89404-170 89404-172 89404-174 89404-176 89404-178 89404-180 89404-182 89404-184 89404-186 89404-188 89404-190 89404-192 89404-194 89404-198 89404-200 89404-202 89404-204 89404-206 89404-208 89404-210 89404-212 89404-214 89404-218 89404-220 89404-222 89404-230 89404-234 89404-236
Tygon S3™ B-44-4X Food and Beverage Dispensing Tubing
Tubing Transfer Tubing
Designed to ensure a bacterial-free fluid path in a wide variety of food processing applications, Tygon S3™ B-44-4X tubing from Saint-Gobain Performance Plastics is now phthalate-free.
Tygon S3™ B-44-4X Food, Milk, and Dairy Tubing features a smooth, non-porous bore that will not trap particulates or promote bacterial growth. Compatible with foods containing a high oil content. Resistant to harsh alkaline cleaners and sanitizers. Excellent alternative to rigid piping systems.
The bio-based Tygon S3™ line combines the high performance standards customers demand with an eco-friendlyMore Product Information