Flip-Top Centrifuge Tubes, Argos Technologies
Supplier: Antyila Scientific
04395-56
89401-198CS 599.74 USD
89401-198
Flip-Top Centrifuge Tubes, Argos Technologies
Tubes Centrifuge Tubes
The Argos flip-top centrifuge tube offers a hinged flip-top cap that allows for single-handed opening and closing.
  • Guaranteed DNase-, RNase-, and pyrogen-free
  • Gamma irradiated
The attached cap provides less chance of dropping or losing a separate cap. Supplied with bold, easy-to-read black graduations that are accurate to ±2%. Tubes feature a large white marking area for sample identification. Manufactured in a class 100000 clean room. Operational temperature range is –80 to 120 °C. Tubes are compatible with any fixed angle or swing bucket rotor.
Order Now
SPECIFICATIONS
Environmentally Preferable