Heated Ultrasonic Baths, Mechanical, Branson Ultrasonics
Supplier: Branson Ultrasonics
CPX-952-117R CPX-952-217R CPX-952-317R CPX-952-517R CPX-952-817R CPX-952-137R CPX-952-237R CPX-952-337R CPX-952-537R CPX-952-837R
89375-470EA 692.32 USD
89375-470 89375-472 89375-474 89375-476 89375-478 89375-480 89375-482 89375-484 89375-486 89375-488
Heated Ultrasonic Baths, Mechanical, Branson Ultrasonics
Ultrasonic Cleaners
MH Series features 60 minute or continuous operation with heating capabilities, offering sweep frequency which eliminates standing waves and creates consistent cavitation through the tank
Transducers are 40KHz rugged industrial transducers. Controls are located above and behind the cleaning tank in an easily accessible control tower panel, and raised above the tank to avoid damage from cleaning solutions. This unit has convenient built-in pour drains on the .5 and .75-gallon models. Tank drains with valves are incorporated on models 1.5-gallon and larger.
Certificaciones: UL, CSA, CE,More Product Information