MAXI FlexTubes, IBI Scientific
Supplier: IBI Scientific
IB48003 IB48006 IB48200 IB48201 IB48202 IB48203 IB48210 IB48220 IB48230 IB48231 IB48232 IB48233 IB48234 IB48235 IB48240 IB48241 IB48242 IB48243 IB48244 IB48245 IB48250 IB48251 IB48252 IB48253 IB48254 IB48255 IB48260 IB48261 IB48262 IB48263 IB48264 IB48265
89369-448EA 29.97 USD
89369-448 89369-454 89369-534 89369-536 89369-538 89369-540 89369-542 89369-544 89369-546 89369-548 89369-550 89369-552 89369-554 89369-556 89369-558 89369-560 89369-562 89369-564 89369-566 89369-568 89369-570 89369-572 89369-574 89369-576 89369-578 89369-580 89369-582 89369-584 89369-586 89369-588 89369-590 89369-592
MAXI FlexTubes, IBI Scientific
Tubes Dialysis Tubes
IBI MAXI FlexTubes are typically used for the extraction of proteins, RNA, DNA, or oligonucleotides (>20 nt) from polyacrylamide, agarose, or any other gel matrix in a running buffer.
MAXI FlexTubes require a minimum of 20 µg of protein at the start of extraction. The maximum size of the gel slice that can be inserted into the MAXI FlexTube is 2×1 cm.
The yield of DNA/RNA recovery from an agarose gel or polyacrylamide gel is 90%. The yield of protein recovery from an SDS-PAGE gel is 70%. The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.
Order Now
SPECIFICATIONS
Environmentally Preferable