Anti-Streptavidin Rabbit Polyclonal Antibody

Supplier: Genetex
GTX27239
89362-504EA 620.21 USD
89362-504
Anti-Streptavidin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR