Anti-SKI Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85522
89329-632EA 620.88 USD
89329-632
Anti-SKI Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Ski

Type: Primary
Antigen: SKI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol SKI
Environmentally Preferable
Antigen Name SKI protein
Antigen Synonyms Ski oncogene|Proto-oncogene c-Ski|SGS|Ski|SKV
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 6497
Isotype
Western Blot Yes
ELISA Yes
ImmunoFluorescence Yes
Purification Affinity chromatography purified via peptide column
Storage Buffer PBS containing 0.02% sodium azide
Storage Temperature Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR