Anti-SLC22A18AS Rabbit Polyclonal Antibody

Supplier: Genetex
GTX119059
89323-624EA 594.42 USD
89323-624
Anti-SLC22A18AS Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SLC22A18AS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol SLC22A18AS
Antigen Name Solute Carrier Family 22 Member A18AS
Environmentally Preferable
Antigen Synonyms BWR1B|p27-BWR1B|BWSCR1B|SLC22A1LS|ORCTL2S
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 5003
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC22A18AS. The exact sequence is proprietary.
Purification Purified by antigen-affinity chromatography.
Concentration 0.75 mg/ml
Storage Buffer 0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
Storage Temperature Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR