Anti-RNF212 Mouse Monoclonal Antibody [clone: INN-hCG-24]

Supplier: Genetex
GTX11390
89317-280EA 619.93 USD
89317-280
Anti-RNF212 Mouse Monoclonal Antibody [clone: INN-hCG-24]
Antibodies
Mouse monoclonal antibody [INN-hCG-24] to CG beta

Type: Primary
Antigen: RNF212
Clonality: Monoclonal
Clone: INN-hCG-24
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR