Anti-CGB Mouse Monoclonal Antibody [clone: 057-10019]

Supplier: Genetex
GTX42560
89299-300EA 639.88 USD
89299-300
Anti-CGB Mouse Monoclonal Antibody [clone: 057-10019]
Antibodies
Mouse Monoclonal antibody [057-10019] to hCG beta

Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: 057-10019
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR