Anti-MBL2 Goat Polyclonal Antibody

Supplier: Genetex
GTX89586
89297-826EA 620.03 USD
89297-826
Anti-MBL2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to MBL2 / Mannan-Binding Lectin

Approx 26kDa band observed in Human Heart and Human Kidney lysates (calculated MW of 26.1kDa according to NP_000233). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: MBL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol MBL2
Environmentally Preferable
Antigen Name Mannan-Binding Lectin 2
Antigen Synonyms MBL2D|MBPD|MBL|HSMBPC|MBP1|MBP|MBP-C|COLEC1
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Gene ID 4153
Isotype IgG
Western Blot Yes
ELISA Yes
Immunogen STSHLAVCEFPI
Purification Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Concentration 0.5mg/ml
Storage Buffer Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Storage Temperature Aliquot and store at –20 °C. Minimize freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR