Anti-CD41 (Integirn alpha 2b (platelet glycoprotein IIb of IIb/IIIa Complex Antigen CD41)) Mouse Monoclonal Antibody [clone: PM6/248]

Supplier: Genetex
GTX76177
89279-970EA 636.6 USD
89279-970
Anti-CD41 (Integirn alpha 2b (platelet glycoprotein IIb of IIb/IIIa Complex Antigen CD41)) Mouse Monoclonal Antibody [clone: PM6/248]
Antibodies
Mouse monoclonal antibody [PM6/248] to CD41

Type: Primary
Antigen: CD41 (Integirn alpha 2b (platelet glycoprotein IIb of IIb/IIIa Complex Antigen CD41))
Clonality: Monoclonal
Clone: PM6/248
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR