Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: B753M]

Supplier: Genetex
GTX39927
89279-330EA 630.87 USD
89279-330
Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: B753M]
Antibodies
Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: Monoclonal
Clone: B753M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR