VWR® Lister Bandage Scissors
Supplier: VWR International
89259-978
89259-978EA 38.86 USD
89259-978
VWR® Lister Bandage Scissors
Scissors
VWR® Lister Bandage Scissors are stainless steel with one blade with a rounded tip and one blade with a blunt, flattened, probe tip.
  • Scissors are angular
Lister bandage scissors are available in a range of lengths and order sizes, from individual to bulk.