Cryostat Blade Carriers, Epredia™, Richard-Allan Scientific
Supplier: Epredia
705460 705470 705580 705840 705940 705950 705630
84000-462EA 4969.14 USD
84000-462 84000-464 84000-470 16002-438 84000-472 89235-302 89235-304 89497-220 10031-420
Cryostat Blade Carriers, Epredia™, Richard-Allan Scientific
Microtomes and Microtome Assemblies Microtome Blades and Handles
Use Epredia™ Cryostat Blade Carriers to match a variety of Epredia™ and Microm™ Cryostats.
Standard Knife Carrier allows for insertion of standard knives, with clearance angle adjustment
Disposable Blade Carrier EC allows for insertion of high- and low-profile blades, with clearance angle adjustment. Can be moved on precision dovetail guide with quick clamp plate. Includes anti-roll plate.
Disposable Blade Carrier EV is for use with vacutome system. Allows for insertion of high- and low-profile disposable blades. Includes a vacuum anti-roll hood for stretching and disposing ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable