Bel-Art Magic Touch 2™ Ice Buckets, ATS Life Sciences
Supplier: Bel-Art Products, a Part of SP
M16807-2001 M16807-2002 M16807-2003 M16807-2004 M16807-4001 M16807-4002 M16807-4003 M16807-4004
89233-340EA 160.94 USD
89233-340 89233-342 89233-344 89233-346 89233-348 89233-350 89233-352 89233-354
Bel-Art Magic Touch 2™ Ice Buckets, ATS Life Sciences
Ice Containers
Superior insulating properties combined with high chemical resistance make Magic Touch 2™ Ice Buckets compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen.
  • Excellent insulation
  • High chemical resistance
  • Convenient pour spout
  • Stackable
  • Lightweight and durable
  • Expanded urethane construction
  • Temperature range: –196...+100 °C (–320... +212 °F)
Able to withstand a wide temperature range, these high-quality, expanded urethane, laboratory ice buckets areMore Product Information