ProGuard® Professional Handcream, Decon Labs
Supplier: Decon Labs
9503 9516
89233-206CS 152.4 USD
89233-206 89233-208
ProGuard® Professional Handcream, Decon Labs
Skincare Lotions Hand Creams
Proguard Professional Handcream combines emollients to moisturize dry, cracked skin with a barrier film that seals in the moisture, seals out the problems
  • Combines emollients to moisturize dry, cracked skin with a barrier film that seals in the moisture, seals out the problems
  • Effective in eliminating the irritation caused by latex gloves
  • Does not contain perfumes, oils, greases, petroleum, aloe or cocoa butter
  • Compatible with and will not compromise the protection of latex gloves
It is particularly effective in eliminating the irritation caused byMore Product Information