Barnstead™ MicroPure Water Purification Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
50132370 50132372 50132373 50132374
89230-424EA 6104.75 USD
89230-424 89230-426 89230-428 89230-430 89230-432 89230-434
Barnstead™ MicroPure Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead MicroPure water purification systems provides up to 15L/day of Type 1, ultrapure (18.2 megohm) water from pre-treated feed water. All MicroPure systems include:feed water monitoring to protect your consumables, quick discconnects to make changing consumables easy, and all core consumables and tubing. MicroPure-ST systems come standard with a 6 liter integrated feed tank for labs that do not have a ready supply of pre-treated feed water.
The MicroPure systems offer highly versatile mounting options.The units can be mounted on the wall, on the bench or under the bench.