VWR® Carboys with Versatile Caps, HDPE
Supplier: VWR International
89170-762 89170-764 89170-766 89170-768 89170-770 89170-772 89170-774 89170-776 89170-778 89170-780 89199-164 89217-552
89170-762EA 188.57 USD
89170-762 89170-764 89170-766 89170-768 89170-770 89170-772 89170-774 89170-776 89170-778 89170-780 89199-164 89217-552
VWR® Carboys with Versatile Caps, HDPE
Bottles Carboys
VWR® HDPE carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
  • Chemical-resistant
  • HDPE construction
  • Molded grips for easy lifting, handling, and pouring
The distinctive rectangular shape saves valuable bench space.
VWR® HDPE carboys are molded from a translucent USP Class VI, and FDA grade resin. They offer excellent chemical resistance, making them ideal for most buffers and reagents as well as powders and solids. Versatile caps, adapters, versatile barbs, and spigots areMore Product Information